Order Entry
Slovenia
ContactUsLinkComponent
Methyl-1-cyclopentene-1-carboxylate
Irritant
Methyl-1-cyclopentene-1-carboxylate
Catalog # APOSOR1990-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-1-cyclopentene-1-carboxylate
Catalog # APOSOR1990-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :126.1531 g/mol
Boiling Pt:76…78 °C
Density:1.031 g/cm³ (25 °C)
Flash Pt:62 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:25662-28-6
Formula:C₇H₁₀O₂
MDL Number:MFCD00239506