
2,3-Dimethoxybenzylamine, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4393-09-3
:
Formula:C₉H₁₃NO₂
MDL Number:MFCD00052392
:167.21
Boiling Pt:137 °C/11mmHg
Density:1.13 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:224-515-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together