
4-tert-Butyl-1-bromobenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3972-65-4
:
Formula:C₁₀H₁₃Br
MDL Number:MFCD00000108
Boiling Pt:106…109 °C/20mmHg
Melting Pt:15…16 °C
Density:1.229 g/cm³ (25 °C)
Flash Pt:97 °C
:213.12 g/mol
EINECS:223-599-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together