
1-Ethynyl-3-nitrobenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3034-94-4
:
Formula:C₈H₅NO₂
MDL Number:MFCD02181150
:147.13 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together