
7-Azaindole-5-boronic acid pinacol ester
Catalog # APOSOR15744-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:244.101 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:754214-56-7
Formula:C₁₃H₁₇BN₂O₂
MDL Number:MFCD08060937