
Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate
Catalog # APOSOR15346-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:218.06 g/mol
Melting Pt:63…65 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1196-90-3
Formula:C₇H₈BrNO₂
MDL Number:MFCD08275077