Order Entry
Slovenia
ContactUsLinkComponent
4-Bromo-1H-pyrrole-2-carboxylic acid 97%
Irritant
4-Bromo-1H-pyrrole-2-carboxylic acid 97%
Catalog # APOSOR15253-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromo-1H-pyrrole-2-carboxylic acid 97%
Catalog # APOSOR15253-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.9948 g/mol
Melting Pt:168…170 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:27746-02-7
Formula:C₅H₄BrNO₂
MDL Number:MFCD00832862