
4H-Furo[3,2-b]pyrrole-5-carboxylic acid methylester 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:77484-99-2
:
Formula:C₈H₇NO₃
MDL Number:MFCD06797487
:165.15 g/mol
Melting Pt:138…140 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together