tert-Butyl-4-(hydrazinocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:187834-88-4
:
Formula:C₁₁H₂₁N₃O₃
MDL Number:MFCD06795927
:243.31 g/mol
Melting Pt:104…106 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together