Order Entry
Slovenia
ContactUsLinkComponent
5-Methylisoxazole-3-carbonitrile 97%
5-Methylisoxazole-3-carbonitrile 97%
Catalog # APOSOR12255-100MG
Supplier:  Apollo Scientific
CAS Number:  
5-Methylisoxazole-3-carbonitrile 97%
Catalog # APOSOR12255-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :108.1 g/mol
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
 :GHS06
CAS Number:57351-99-2
Formula:C₅H₄N₂O
MDL Number:MFCD09064940