0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Methyl-4-(3-bromopropoxy)benzenecarboxylate
Methyl-4-(3-bromopropoxy)benzenecarboxylate
Methyl-4-(3-bromopropoxy)benzenecarboxylate
APOSOR12129-1G
:
Apollo Scientific
:
Methyl-4-(3-bromopropoxy)benzenecarboxylate
APOSOR12129-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
273.13 g/mol
:
71…73 °C
:
GHS07
:
Ambient
:
135998-88-8
:
C₁₁H₁₃BrO₃
:
MFCD01814575