2-Methoxy-5-(methoxycarbonyl)phenylboronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:221006-63-9
:
Formula:C₉H₁₁BO₅
MDL Number:MFCD09800877
:209.9916 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together