
3-Chloro-5-(4-morpholinylcarbonyl)phenylboronic acid
Catalog # APOSOR11656-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
Melting Pt:143…146 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:957120-55-7
Formula:C₁₁H₁₃BClNO₄
MDL Number:MFCD09800860