2,3-Dimethoxyphenylboronic acid
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2,3-Dimethoxybenzeneboronic acid, (2,3-Dimethoxyphenyl)metaboric acid, Dihydroxy(2,3-dimethoxyphenyl)borane
CAS Number:40972-86-9
:
Formula:C₈H₁₁BO₄
MDL Number:MFCD02683112
Molecular Weight:181.98 g/mol
Melting Pt:67…71 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together