
4-(3-Bromophenyl)-2-butanone
Catalog # APOSOR110679-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.11 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:3506-70-5
Formula:C₁₀H₁₁BrO
MDL Number:MFCD09991820