0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
5-Indoleboronic acid pinacol ester
5-Indoleboronic acid pinacol ester
5-Indoleboronic acid pinacol ester
APOSOR11011-5G
:
Apollo Scientific
:
5-Indoleboronic acid pinacol ester
APOSOR11011-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
243.1092 g/mol
:
114…125 °C
:
GHS07
:
Ambient
:
269410-24-4
:
C₁₄H₁₈BNO₂
:
MFCD03789263