
Ethyl-2-bromothiazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:100367-77-9
:
Formula:C₆H₆BrNO₂S
MDL Number:MFCD03788564
Melting Pt:68…70 °C
Flash Pt:>110 °C
:236.08 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together