Ethyl-3-chloroimidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000017-95-7
:
Formula:C₁₀H₉ClN₂O₂
MDL Number:MFCD09842616
:224.65 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together