Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole 97%
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole 97%
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole 97%
APOSOR01543-50MG
:
Apollo Scientific
:
5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole 97%
APOSOR01543-50MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
50 mg
Seq:
2370
:
:
161.16 g/mol
:
GHS07
:
Ambient
:
10350-70-6
:
C₈H₇N₃O
:
MFCD00464110