

α,α'-Dibromo-p-xylene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:623-24-5
:
Formula:C₈H₈Br₂
MDL Number:MFCD00000182
:263.96 g/mol
Boiling Pt:156 °C/23mmHg
Melting Pt:145…147 °C
Density:1.51 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:210-781-1
UN:3448
ADR:6.1,II
:GHS05
:GHS07
Frequently Bought Together