0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Magnesium formate dihydrate
Magnesium formate dihydrate 98+%
Magnesium formate dihydrate 98+%
APOSIN2449-100G
:
Apollo Scientific
:
Magnesium formate dihydrate 98+%
APOSIN2449-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
150.37 g/mol
:
GHS07
:
Ambient
:
6150-82-9
:
C₂H₂MgO₄·2H₂O
:
MFCD00150512