Order Entry
Slovenia
ContactUsLinkComponent
Myriocin (from Mycelia sterilia)
Irritant
Myriocin (from Mycelia sterilia)
Catalog # APOSBIM1005-10MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Myriocin (from Mycelia sterilia)
Catalog # APOSBIM1005-10MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
Melting Pt:175…180 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:35891-70-4
MDL Number:MFCD01632772