EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid) ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16052-06-5
:
Formula:C₉H₂₀N₂O₄S
MDL Number:MFCD00006160
Melting Pt:237…239 °C
:252.33 g/mol
EINECS:240-198-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together