N-Acetyl-L-alanine
Catalog # APOSBIA1219-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:131.131 g/mol
Melting Pt:125…126 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:97-69-8
Formula:C₅H₉NO₃
MDL Number:MFCD00063132