Order Entry
Slovenia
ContactUsLinkComponent
Mouse recombinant OX40 ligand (from HEK293)
Mouse recombinant OX40 ligand (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Mouse recombinant OX40 ligand (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Gene ID:
    P43488
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    TNFSF4|CD134L|TXGP1|OX40L|Glycoprotein Gp34|CD134 ligand|CD252
  • Protein/peptide name:
    OX40 Ligand
  • Purity:
    >95%
  • Molecular weight:
    43,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together