Human recombinant LAG-3 (from HEK293 cells)
Catalog # ACRBLA3-H522A100UG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,01 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Anti-Human LAG3 MAb, Human IgG1 at 2 μg/ml (100 μl/well) can bind Human LAG-3, His Tag, low endotoxin with a linear range of 1 to 6 ng/ml (QC tested).
- Gene ID:P18627-1
- Protein synonyms:FDC|CD223|LAG3
- Protein/peptide name:LAG-3
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:47,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT