Cynomolgus monkey recombinant PSMA (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Tag sequence:His Tag (MALS verified) (active enzyme)
- Storage conditions:−70 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Gene ID:A0A2K5VNZ0-1
- Protein synonyms:FOLH|FOLH1|PSM|GCPII|NAALAD1|PSMA|FGCP|NAALADase I|GIG27
- Protein/peptide name:PSMA
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:81,4 kDa
- Endotoxin level:Low
- Formulation:Delivered as bulk protein in a 0,2 μm filtered solution of MES and NaCl with trehalose as protectant.
- Shipping temperature:Dry Ice
Specifications
Frequently Bought Together