
Methyl 3-bromo-5-fluoropyridine-2-carboxylate
Catalog # APOSPC57016-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:234.023 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1214337-00-4
Formula:C₇H₅BrFNO₂
MDL Number:MFCD14698117