
4-Amino-3-(4-fluorophenyl)butyric acid methyl ester hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2149591-12-6
:
Formula:C₁₁H₁₄FNO₂·ClH
MDL Number:MFCD30492328
:247.694 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together