2-Amino-3-(trifluoromethoxy)propionic acid hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1301738-59-9
:
Formula:C₄H₆F₃NO₃
MDL Number:MFCD17677130
:173.091 g/mol
Storage Temperature:Ambient
Frequently Bought Together