4-Cyclopropyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid ethyl ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:887409-11-2
:
Formula:C₁₁H₁₁F₃N₂O₂
MDL Number:MFCD07367384
:260.212 g/mol
Storage Temperature:Ambient
Frequently Bought Together