2,3-Dimethyl-5-fluoroindole
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 5-Fluoro-2,3-dimethyl-1H-indole
CAS Number:526-47-6
:
Formula:C₁₀H₁₀FN
MDL Number:MFCD01158420
Molecular Weight:163.192 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together