5-(Difluoromethyl)-4-methyl-thiophene-2-carboxylic acid
Catalog # APOSPC520214-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.183 g/mol
Storage Temperature:Ambient
CAS Number:2092062-21-8
Formula:C₇H₆F₂O₂S
MDL Number:MFCD29767694