Methyl 6-bromo-3-(difluoromethyl)-1H-indazole-4-carboxylate
Catalog # APOSPC520143-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:305.076 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₇BrF₂N₂O₂
MDL Number:MFCD29767626