Order Entry
Slovenia
ContactUsLinkComponent
3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
Catalog # APOSPC510272-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
Catalog # APOSPC510272-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.184 g/mol
Storage Temperature:Ambient
CAS Number:2006277-43-4
Formula:C₁₁H₈FNO₄
MDL Number:MFCD28977389