3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylic acid
Catalog # APOSPC510268-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:287.191 g/mol
Storage Temperature:Ambient
CAS Number:2006277-73-0
Formula:C₁₂H₈F₃NO₄
MDL Number:MFCD28977397