3-[[(2-Tetrahydropyranyl)oxy][4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylic acid
Catalog # APOSPC510267-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:371.308 g/mol
Storage Temperature:Ambient
CAS Number:2006277-46-7
Formula:C₁₇H₁₆F₃NO₅
MDL Number:MFCD28977398