4,5-Difluoro-2-(hydroxymethyl)benzimidazole
Catalog # APOSPC510254-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.143 g/mol
Storage Temperature:Ambient
CAS Number:1517146-73-4
Formula:C₈H₆F₂N₂O
MDL Number:MFCD26034283