Order Entry
Slovenia
ContactUsLinkComponent
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methyl-5-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methyl-5-nitrophenyl ether 95%
Catalog # APOSPC502754-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methyl-5-nitrophenyl ether 95%
Catalog # APOSPC502754-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-2-fluoro-4-methyl-5-nitrobenzene
 :
 :351.201 g/mol
Storage Temperature:Ambient
CAS Number:2244085-94-5
Formula:C₁₄H₇F₆NO₃
MDL Number:MFCD31630035