Order Entry
Slovenia
ContactUsLinkComponent
4-Bromo-3,3,4,4-tetrafluorobutyl ethyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl ethyl carbonate
Catalog # APOSPC450417-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3,3,4,4-tetrafluorobutyl ethyl carbonate
Catalog # APOSPC450417-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :297.042 g/mol
Storage Temperature:Ambient
CAS Number:1923065-49-9
Formula:C₇H₉BrF₄O₃
MDL Number:MFCD28334470