Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 1H,1H-perfluorohexyl carbonate
Ethyl 1H,1H-perfluorohexyl carbonate
Ethyl 1H,1H-perfluorohexyl carbonate
APOSPC450413-25G
:
Apollo Scientific
:
Ethyl 1H,1H-perfluorohexyl carbonate
APOSPC450413-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
372.133 g/mol
:
Ambient
:
1980034-41-0
:
C₉H₇F₁₁O₃
:
MFCD28334390