Ethyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Catalog # APOSPC450411-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:254.127 g/mol
Storage Temperature:Ambient
CAS Number:1035454-56-8
Formula:C₇H₈F₆O₃
MDL Number:MFCD28334388