Ethyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Catalog # APOSPC450410-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:272.118 g/mol
Storage Temperature:Ambient
CAS Number:1980034-03-4
Formula:C₇H₇F₇O₃
MDL Number:MFCD28334387