3-Methyl-5-(4-trifluoromethyl-phenyl)-isoxazole-4-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:220652-98-2
:
Formula:C₁₂H₈F₃NO₃
MDL Number:MFCD11042908
:271.192 g/mol
Storage Temperature:Ambient
Frequently Bought Together