N-Allyl-2-(3-(trifluoromethyl)-4,5,6,7-tetrahydroindazol-1-yl)acetamide
Catalog # APOSPC430601-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:287.281 g/mol
Storage Temperature:Ambient
CAS Number:1258652-68-4
Formula:C₁₃H₁₆F₃N₃O
MDL Number:MFCD18157704