1-[(2-Fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410357-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:236.199 g/mol
Storage Temperature:Ambient
CAS Number:1005662-40-7
Formula:C₁₁H₉FN₂O₃
MDL Number:MFCD04050603