1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410311-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.207 g/mol
Storage Temperature:Ambient
CAS Number:1004193-05-8
Formula:C₁₂H₉F₃N₂O₃
MDL Number:MFCD04967890