Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-3-chloro-8-fluoroquinoline
4-Bromo-3-chloro-8-fluoroquinoline
4-Bromo-3-chloro-8-fluoroquinoline
APOSPC404604-1G
:
Apollo Scientific
:
4-Bromo-3-chloro-8-fluoroquinoline
APOSPC404604-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
260.49 g/mol
:
Ambient
:
1211659-63-0
:
C₉H₄BrClFN
:
MFCD13619702