2,3-Difluoro-4-methoxyphenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886503-74-8
:
Formula:C₉H₇F₂NO
MDL Number:MFCD06660320
:183.155 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
Frequently Bought Together