
3-(4-Bromophenyl)-5-(trifluoromethyl)isoxazole-4-carboxylic acid
Catalog # APOSPC200553-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:336.062 g/mol
Melting Pt:145…148 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1159978-82-1
Formula:C₁₁H₅BrF₃NO₃
MDL Number:MFCD12196379